missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Cxcl5 (Mouse) ELISA Kit

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Mouse Cxcl5

Brand:  Abnova™ KA1799.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006835

  • 578.00 EUR / 1 set


Estimated Shipment
11-02-2020
Add to basket

Specifications

Specifications

Biotin
Cxcl5 (Mouse) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse Cxcl5.
Colorimetric
96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP Streptavidian concentrate, 12mL TMB One-Step Substrate reagent, 8mL stop solut
96 Assays
Cell Culture Supernatant, Plasma, Serum
96 Assays
Mouse
35pg/mL
Quantification
Cxcl5 (Mouse) ELISA Kit
RUO
Store the kit at -20°C. Standard should be stored at -20°C or -80°C. Avoid repeated freezing and thawing.
100μL
Videos
Documents