missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Cxcl5 (Mouse) ELISA Kit
In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Mouse Cxcl5
POA
Specifications
Detection Method | Colorimetric |
---|---|
Cross Reactivity | Mouse |
Product Type | Cxcl5 (Mouse) ELISA Kit |
Quantity | 96 Assays |
Regulatory Status | For research use only (RUO) |
Specifications
Colorimetric | |
Cxcl5 (Mouse) ELISA Kit | |
For research use only (RUO) |
Mouse | |
96 Assays | |
100 μL |