Learn More
Abnova™ CXCL10 (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human CXCL10
Brand:  Abnova™ KA2004.1Kit
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16006995
  • 578.00 EUR / 1 set
Estimated Shipment
19-03-2020
Add to basket
Specifications
Specifications
31.2 to 2000pg/mL
Human
<1pg/mL
Quantification
CXCL10 (Human) ELISA Kit
1 Kit
Store the kit at-20°C.
Biotin
CXCL10 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human CXCL10.
Colorimetric
96-well Antibody coated microwell, 10ng x 2 vials lyophilized recombinant Human CXCL 10 standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Human CXCL 10 Antibody, 12mL Antibody Diluent Buffer, 130μL Avidin-Bi
CXCL10 (Human) ELISA kit
Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
100μL