missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CTSB (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human CTSB
POA
Specifications
Calibration Range | 156 to 10000pg/mL |
---|---|
Detection Limit | 5pg/mL |
Detection Method | Colorimetric |
Conjugate | Biotin |
Cross Reactivity | Human |
Specifications
156 to 10000pg/mL | |
Colorimetric | |
Human | |
Quantification | |
96 Assays |
5pg/mL | |
Biotin | |
CTSB (Human) ELISA Kit | |
96-well plate pre-coated with anti-Human Cathepsin B antibody, Lyophilized Human Cathepsin B standards: 10ng x 2, Sample / Standard diluent buffer: 30mL, Biotin conjugated anti-Human Cathepsin B antibody (Concentrated Dilution: 1:100): 130uL, Antibody diluent buffer: 12mL, Avidin-Biotin-Peroxidase Complex (ABC) (ConcentratedDilution: 1:100): 130uL, ABC diluent buffer: 12mL, TMB substrate: 10mL, Stop solution: 10mL, Wash buffer (25X): 30mL | |
100 μL |