missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CTSB (Human) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Human CTSB

Brand:  Abnova™ KA3024.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16007775

  • 578.00 EUR / 1 set


Estimated Shipment
12-02-2020
Add to basket

Specifications

Specifications

156 to 10000pg/mL
Human
5pg/mL
Quantification
CTSB (Human) ELISA Kit
Body Fluid, Cell Culture Supernate, Plasma, Serum, Tissue Lysate
96 Assays
Biotin
CTSB (Human) ELISA Kit is intended for the quantitative measurement of human CTSB.
Colorimetric
96-well plate pre-coated with anti-Human Cathepsin B antibody, Lyophilized Human Cathepsin B standards: 10ng x 2, Sample / Standard diluent buffer: 30mL, Biotin conjugated anti-Human Cathepsin B antibody (Concentrated Dilution: 1:100): 130uL, Antibody diluent buffer: 12mL, Avidin-Biotin-Peroxidase Complex (ABC) (ConcentratedDilution: 1:100): 130uL, ABC diluent buffer: 12mL, TMB substrate: 10mL, Stop solution: 10mL, Wash buffer (25X): 30mL
96 Assays
Store at 4°C for 4 months, or at -20°C for 8 months.
100μL
Videos
Documents