missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Cortisol (Sheep) ELISA Kit

Immunoassay for the quantitative determination of cortisol in sheep serum, plasma, urine

Brand:  Abnova™ KA0919.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006405

  • 578.00 EUR / 1 set


Estimated Shipment
20-02-2020
Add to basket

Specifications

Specifications

1 to 80ng/mL
Sheep
0.1ng/mL
Quantification
Cortisol (Sheep) ELISA Kit
RUO
Store the kit at 4°C.
HRP
Cortisol (Sheep) ELISA Kit is used for the quantitative measurement of Cortisol in sheep serum or plasma.
Colorimetric
12 x 8 x 1 Microwell coated with Cortisol MAb, 0.7mL Cortisol standard, 1.2mL (20X) Enzyme Conjugate, 12mL TMB Substrate, 12mL stop solution, 25mL (20X) Wash concentrate, 24mL Assay diluent
96 Assays
Plasma, Serum
40μL
Videos
Documents