missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Chloramphenicol ELISA Kit

Competitive binding Enzyme Immunoassay for the quantitative measurement of Chloramphenicol

Brand:  Abnova™ KA1415.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006635

  • 263.00 EUR / 1 set


Estimated Shipment
19-02-2020
Add to basket

Specifications

Specifications

AP
Colorimetric
96-well microtiter plate Twelve strip of 8 detachable well, 1.2mL negative calibrator, 3 vials positive calibrator, 10.5mL Chloramphenicol Alkaline Phosphatase Conjugate (CPC ALP),, p-Nitrophenyl Phosphate (pNPP)Substrate,15mL Wash Buffer,5.5mL stop solution
Chloramphenicol ELISA kit
50μL
50μL
Chloramphenicol ELISA Kit is a competitive binding enzyme immunoassay for the quantitative measurement of chloramphenicol.
Quantification
Chloramphenicol ELISA Kit
1 Kit
Store the kit at 4°C.
Videos
Documents