missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CD40 (Human) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Human CD40

Brand:  Abnova™ KA1061.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006515

  • 578.00 EUR / 1 set


Estimated Shipment
03-09-2020
Add to basket

Specifications

Specifications

15.6 to 1000pg/mL
Human
1pg/mL
Quantification
CD40 (Human) ELISA Kit
1 Kit
Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Serum, Saliva, Urine
Store the kit at-20°C.
100μL
Biotin
Human CD40 ELISA kit is a sandwich enzyme immunoassay for the quantitative measurement of human CD40.
Colorimetric
96-well Antibody coated microwell, 10ng x 2 vials lyophilized recombinant Human CD40 standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Human CD40 Antibody,, 12mL Antibody Diluent Buffer,130μL Avidin Biotin Peroxidase Complex,12mL ABC diluent buffer, 10mL TMB color developing agent, 10mL TMB stop solution
CD40 (Human) ELISA kit
RUO
100μL
96 Assays
Videos
Documents