missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CD40 (Human) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Human CD40

Brand:  Abnova™ KA0161.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16005905

  • 578.00 EUR / 1 set


Estimated Shipment
11-02-2020
Add to basket

Specifications

Specifications

7.8 to 500pg/mL
Human
1.3pg/mL
Quantification
CD40 (Human) ELISA Kit
1 Kit
Amniotic Fluid, Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Store the controls at -20°C. Others at 4°C. Avoid repeated freezing and thawing.
50μL
Biotin
CD40 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human sCD40.
Colorimetric
96 wells-Antibody coated microwells, Biotin-Conjugate antibody: 100μL, Streptavidin-HRP: 150μL, Standard (lyophilized): 2 vials, Sample Diluent: 12mL, Assay Buffer Concentrate 20X: 5mL, Wash Buffer Concentrate 20X: 50 mL, Substrate Solution: 15mL, Stop Solution: 15mL, Blue-Dye: 0.4mL, Green-Dye: 0.4mL, Red-Dye: 0.4mL
CD40 (Human) ELISA kit
RUO
50μL
96 Assays
Videos
Documents