missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CCL8 (Human) ELISA Kit

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human CCL8

Brand:  Abnova™ KA3084.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16007825

  • 578.00 EUR / 1 set


Estimated Shipment
13-03-2020
Add to basket

Specifications

Specifications

30 to 1000pg/mL
Human
10pg/mL
Quantification
CCL8 (Human) ELISA Kit
RUO
Store at 4°C for 6 months.
10μL
HRP
CCL8 (Human) ELISA Kit is intended for the quantitative measurement of human CCL8.
Colorimetric
96-well plate pre-coated with anti-human MCP-2antibody, Standard (0.5mL), Standard diluent buffer (1.5mL), 30X wash buffer (20mL), Sample diluent buffer (6mL), HRP conjugated anti-human MCP-2 antibody (6mL), Stop solution (6mL), TMB Substrate A(6mL), TMB Substrate B (6mL), Plate sealer, Hermetic bag
96 Assays
Cell Culture Supernatant, Plasma, Serum, Tissue Sample, Urine
96 Assays
Videos
Documents