missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Cardiolipin IgM ELISA Kit

Detection of IgM antibody to Cardiolipin in Human serum or plasma.

Brand:  Abnova™ KA0942.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006425

  • 263.00 EUR / 1 set


Estimated Shipment
31-07-2020
Add to basket

Specifications

Specifications

HRP
Colorimetric
12 x 8 x 1 Microwell coated with Cardiolipin Antigen, 22mL sample diluent, 1.5mL calibrator, 1.5mL positive control 1 vial, 1.5mL negative control, 12mL Enzyme Conjugate,12mL TMB Substrate,12mL stop solution,25mL 20X wash concentrate
96 Assays
Store the kit at 4°C.
10μL
Cardiolipin IgM ELISA Kit is used for the detection of IgM antibody to Cardiolipin in human serum or plasma.
Qualitative
Cardiolipin IgM ELISA Kit
Plasma, Serum
96 Assays
Videos
Documents