missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ AHSG (Human) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Human AHSG

Brand:  Abnova™ KA1071.1Kit

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16006525

  • 578.00 EUR / 1 set


Estimated Shipment
31-07-2020
Add to basket

Specifications

Specifications

0.312 to 20ng/mL
Human
<10 pg/mL
Quantification
AHSG (Human) ELISA Kit
1 Kit
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
96 Assays
Biotin
Human AHSG ELISA kit is a sandwich enzyme immunoassay for the quantitative measurement of human AHSG.
Colorimetric
96-well Antibody coated microwell plate, 10ng x 2 vials lyophilized recombinant Human AGER standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Human AGER Antibody, 12mL Antibody Diluent Buffer, 130μL avidin-bi
AHSG (Human) ELISA kit
RUO
Store the kit at-20°C.
100μL
Videos
Documents