Learn More
CleanSpace Technology™ Combined Filter
Combined filter suitable for protection against airborne particulate (dust, mists, and fumes) and organic gases or vapors (boiling point > 65°C).
Brand: CleanSpace Technology™ PAF-0051
Additional Details : Weight : 0.23550kg
Description
- Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
- Materials: Fibreglass particulate media, activated carbon and plastic casing, silicone seal
- Easily fitted and removed from the power unit
Specifications
EN 12942 | |
Filter | |
CleanSpace PAPR Power Units | |
TM3 ABE1 P SL R |