Learn More
Uvex™ 9770 Pheos E-WR Unventilated Safety Helmets
Lightweight, unventilated helmets in a sporty design ideal for use in the electrician sector with low voltage systems. Features interior equipment with a rotating wheel system for strapless width adjustment.
18.10 EUR - 1030.00 EUR
Specifications
Certifications/Compliance | EN 397:2012, EN 50365:2002 |
---|---|
For Use With (Application) | Low Voltage Systems |
Includes | Adapter System for Uvex™ Visors, Helmet Capsules, and Helmet Lamps |
Material | HDPE |
Model | 9770 Pheos E-WR |
Product Code | Brand | Color | Finish | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Finish | Price | Quantity & Availability | ||||
17259111
|
Uvex™
9770536 |
Blue | Reflective |
41.00 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
17239111
|
Uvex™
9770530 |
Blue | Non-reflective |
18.10 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
17269111
|
Uvex™
9770830 |
Gray | Non-reflective |
18.10 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
17299101
|
Uvex™
9770430 |
Green | Non-reflective |
18.10 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
17239101
|
Uvex™
9770230 |
Orange | Non-reflective |
18.10 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
17279101
|
Uvex™
9770330 |
Red | Non-reflective |
18.10 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
17269091
|
Uvex™
9770036 |
White | Reflective |
1030.00 EUR
Pack of 25 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
17209101
|
Uvex™
9770136 |
Yellow | Reflective |
1030.00 EUR
Pack of 25 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
Description
- Lightweight, unventilated helmet in a sporty design
- Completely closed for use in the electrician sector with low voltage systems
- Includes an adapter system for visors, helmet caps, and helmet lamps
- Interior equipment with a rotating wheel system for stepless width adjustment
- Reflective helmets are supplied with Reflex Set L pre-attached to increase worker visibility and safety
Specifications
EN 397:2012, EN 50365:2002 | |
Adapter System for Uvex™ Visors, Helmet Capsules, and Helmet Lamps | |
9770 Pheos E-WR | |
52 to 61 cm |
Low Voltage Systems | |
HDPE | |
Electrically Insulating Helmet | |
Not Ventilated |