Learn More
8-Azahypoxanthine, 98%, Thermo Scientific Chemicals
Catalog Number p-61600
Click to view available options
Quantity:
1 g
5 g
2683-90-1
C4H3N5O
137.10
MFCD00005804
OEEYCNOOAHGFHL-UHFFFAOYSA-N
8-azahypoxanthine, azahypoxanthine, 3h-1,2,3 triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-1,2,3,4,6-pentaazaindene, 7h-1,2,3-triazolo 4,5-d pyrimidin-7-one, 1,4-dihydro, 7-hydroxy-v-triazolo d pyrimidine, 1h-1,2,3-triazolo 4,5-d pyrimidin-7-ol, v-triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-3h-1,2,3 triazolo 4,5-d pyrimidine
75895
2,3-dihydrotriazolo[4,5-d]pyrimidin-7-one
O=C1N=CN=C2NNN=C12
2683-90-1
C4H3N5O
137.10
MFCD00005804
OEEYCNOOAHGFHL-UHFFFAOYSA-N
8-azahypoxanthine, azahypoxanthine, 3h-1,2,3 triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-1,2,3,4,6-pentaazaindene, 7h-1,2,3-triazolo 4,5-d pyrimidin-7-one, 1,4-dihydro, 7-hydroxy-v-triazolo d pyrimidine, 1h-1,2,3-triazolo 4,5-d pyrimidin-7-ol, v-triazolo 4,5-d pyrimidin-7-ol, 7-hydroxy-3h-1,2,3 triazolo 4,5-d pyrimidine
75895
2,3-dihydrotriazolo[4,5-d]pyrimidin-7-one
O=C1N=CN=C2NNN=C12
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 26, IV, 4130
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkaline solution at ph >9
Formula Weight 137.1
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 8-Azahypoxanthine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 8-Azahypoxanthine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.