missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline, 98%, Thermo Scientific™

Catalog Number p-2690927
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4602-73-7
C10H11NO2
177.2
MFCD00143517
UQBWYWCIBNWMPI-UHFFFAOYSA-N
7-hydroxy-6-methoxy-3,4-dihydroisoquinoline, 6-methoxy-3,4-dihydro-7-isoquinolinol, 6-methoxy-7-hydroxy-3,4-dihydroisoquinoline, 3,4-dihydro-7-hydroxy-6-methoxyisoquinoline, 6-methoxy-7-hydroxy-3,4-dihydro-isoquinoline, 7-isoquinolinol, 3,4-dihydro-6-methoxy, pubchem12735, ksc496q9t, 7-hydroxy-6-methoxy-3?4-dihydroisoquinoline, 3,4-dihydro-7-hydroxy-6-methoxy-isoquinoline
363043
6-methoxy-3,4-dihydroisoquinolin-7-ol
COC1=C(C=C2C=NCCC2=C1)O
This item is not returnable. View return policy
4602-73-7
C10H11NO2
177.2
MFCD00143517
UQBWYWCIBNWMPI-UHFFFAOYSA-N
7-hydroxy-6-methoxy-3,4-dihydroisoquinoline, 6-methoxy-3,4-dihydro-7-isoquinolinol, 6-methoxy-7-hydroxy-3,4-dihydroisoquinoline, 3,4-dihydro-7-hydroxy-6-methoxyisoquinoline, 6-methoxy-7-hydroxy-3,4-dihydro-isoquinoline, 7-isoquinolinol, 3,4-dihydro-6-methoxy, pubchem12735, ksc496q9t, 7-hydroxy-6-methoxy-3?4-dihydroisoquinoline, 3,4-dihydro-7-hydroxy-6-methoxy-isoquinoline
363043
6-methoxy-3,4-dihydroisoquinolin-7-ol
COC1=C(C=C2C=NCCC2=C1)O
CAS Min % 97.5
CAS Max % 100.0
Melting Point 187°C to 191°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in chloroform,ethanol,methanol and hot,ethyl acetate
Formula Weight 177.2
Percent Purity 98%
Chemical Name or Material 7-Hydroxy-6-methoxy-3, 4-dihydroisoquinoline, 98%
compliance-icons
Product Identifier
  • 7-Hydroxy-6-methoxy-3,4-dihydroisoquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'