missing translation for 'onlineSavingsMsg'
Learn More

7-Ethyl-10-hydroxycamptothecin, 98%, Thermo Scientific Chemicals

Catalog Number p-7077775
Click to view available options
Quantity:
100 mg
1 g
This item is not returnable. View return policy
86639-52-3
C22H20N2O5
392.4
FJHBVJOVLFPMQE-QFIPXVFZSA-N
7-ethyl-10-hydroxycamptothecin, sn 38 lactone, 7-ethyl-10-hydroxy-camptothecin, 10-hydroxy-7-ethylcamptothecin, 7-ethyl-10-hydroxycamptothecine, le-sn38, 7-ethyl-10-hydroxy-20 s-camptothecin, captothecin, 7-ethyl-10-hydroxy, s-4,11-diethyl-4,9-dihydroxy-1h-pyrano 3',4':6,7 indolizino 1,2-b quinoline-3,14 4h,12h-dione, 7-ethyl-10-hydroxy camptothecin
104842
CHEBI:8988
CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)O
This item is not returnable. View return policy
86639-52-3
C22H20N2O5
392.4
FJHBVJOVLFPMQE-QFIPXVFZSA-N
7-ethyl-10-hydroxycamptothecin, sn 38 lactone, 7-ethyl-10-hydroxy-camptothecin, 10-hydroxy-7-ethylcamptothecin, 7-ethyl-10-hydroxycamptothecine, le-sn38, 7-ethyl-10-hydroxy-20 s-camptothecin, captothecin, 7-ethyl-10-hydroxy, s-4,11-diethyl-4,9-dihydroxy-1h-pyrano 3',4':6,7 indolizino 1,2-b quinoline-3,14 4h,12h-dione, 7-ethyl-10-hydroxy camptothecin
104842
CHEBI:8988
CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 mg
Formula Weight 392.4
Percent Purity 98%
Chemical Name or Material 7-Ethyl-10-hydroxycamptothecin
compliance-icons
Product Identifier
  • 7-Ethyl-10-hydroxycamptothecin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'