missing translation for 'onlineSavingsMsg'
Learn More

7-Deazaguanine, 90+%, Acros Organics

Brand:  Acros Organics 450260050

Additional Details : CAS Number : 7355-55-7 Weight : 0.00500kg


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 15254814

  • 166.20 EUR / 5g


Estimated Shipment date
from Supplier 09-07-2019
Add to basket


Specifications

Chemical Identifiers

7355-55-7
150.141
7-deazaguanine, 2-amino-3,7-dihydropyrrolo 2,3-d pyrimidin-4-one, 2-amino-7h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-4-hydroxypyrrolo 2,3-d pyrimidine, 2-amino-3h-pyrrolo 2,3-d pyrimidin-4 7h-one, unii-gpl8t5zo3m, 2-amino-3,7-dihydro-pyrrolo 2,3-d pyrimidin-4-one, gpl8t5zo3m, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one
2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
C6H6N4O
OLAFFPNXVJANFR-UHFFFAOYSA-N
96253
C1=CNC2=C1C(=O)N=C(N2)N

Specifications

7-Deazaguanine
>230.0°C
7355-55-7
C6H6N4O
OLAFFPNXVJANFR-UHFFFAOYSA-N
2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
96253
90+%
Glass bottle
5g
90+%
7-deazaguanine, 2-amino-3,7-dihydropyrrolo 2,3-d pyrimidin-4-one, 2-amino-7h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-4-hydroxypyrrolo 2,3-d pyrimidine, 2-amino-3h-pyrrolo 2,3-d pyrimidin-4 7h-one, unii-gpl8t5zo3m, 2-amino-3,7-dihydro-pyrrolo 2,3-d pyrimidin-4-one, gpl8t5zo3m, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one
C1=CNC2=C1C(=O)N=C(N2)N
150.141
150.14
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 7-Deazaguanine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documents