Learn More
6-Nitropiperonal, 98+%, ACROS Organics™
Brand:  Acros Organics 160070250
UNSPSC : 12350000
Additional Details : CAS Number : 712-97-0 Weight : 0.07500kg
Quantity 25g
 View more versions of this product
Product Code. 10011930
  • 79.40 EUR / 25g
Estimated Shipment date
from Supplier 28-08-2017
Add to basket
Specifications
Chemical Identifiers
6-Nitropiperonal
712-97-0
100.0
98+%
195.13
98+%
98.0
≥98%
C8H5NO5
MFCD00005829
Specifications
6-Nitropiperonal
712-97-0
100.0
Glass bottle
≥98%
C8H5NO5
195.13
92.0° to 98.0°C
98+%
98.0
Authentic
Fine Crystalline Powder
98+%
Yellow
MFCD00005829
25g
Safety and Handling
Safety and Handling
Product Identifier
  • 6-Nitropiperonal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Documentation
Technical Documents