Learn More
6-Methylpyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Catalog Number 15264348
Click to view available options
Quantity:
250 mg
1 g
610768-32-6
C12H18BNO2
219.09
MFCD08669608
JEECGGXGHJUYMN-UHFFFAOYSA-N
2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-picoline-5-boronic acid pinacol ester, 6-methylpyridine-3-boronic acid pinacol ester, 2-picoline-5-boronic acid pinacolate, 2-methylpyridine-5-boronic acid, pinacol ester, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, pyridine, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-picoline-5-boronic acidpinacolate, 2-methylpyridine-5-boronic acid pinacolate
16414272
2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1
610768-32-6
C12H18BNO2
219.09
MFCD08669608
JEECGGXGHJUYMN-UHFFFAOYSA-N
2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-picoline-5-boronic acid pinacol ester, 6-methylpyridine-3-boronic acid pinacol ester, 2-picoline-5-boronic acid pinacolate, 2-methylpyridine-5-boronic acid, pinacol ester, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, pyridine, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-picoline-5-boronic acidpinacolate, 2-methylpyridine-5-boronic acid pinacolate
16414272
2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 250 mg
Formula Weight 219.09
Percent Purity 95%
Chemical Name or Material 6-Methylpyridine-3-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 6-Methylpyridine-3-boronic acid pinacol ester, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.