missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoropyridine-3-carboxaldehyde, 95%, Acros Organics

Brand:  Acros Organics 449850050

Additional Details : CAS Number : 677728-92-6 Weight : 0.00500kg


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 15254784

  • 195.80 EUR / 5g


In Stock 3
Add to basket


Specifications

Chemical Identifiers

677728-92-6
125.1
2-fluoropyridine-5-carbaldehyde, 6-fluoronicotinaldehyde, 2-fluoro-5-formylpyridine, 2-fluoropyridine-5-carboxaldehyde, 6-fluoro-3-pyridinecarboxaldehyde, 3-pyridinecarboxaldehyde, 6-fluoro, 6-fluoropyridine-3-carboxaldehyde, pubchem5150, zlchem 1162, acmc-1b2mv
6-fluoropyridine-3-carbaldehyde
C6H4FNO
PZPNGWWKCSJKOS-UHFFFAOYSA-N
16414246
C1=CC(=NC=C1C=O)F

Specifications

6-Fluoropyridine-3-carboxaldehyde
5g
95%
2-fluoropyridine-5-carbaldehyde, 6-fluoronicotinaldehyde, 2-fluoro-5-formylpyridine, 2-fluoropyridine-5-carboxaldehyde, 6-fluoro-3-pyridinecarboxaldehyde, 3-pyridinecarboxaldehyde, 6-fluoro, 6-fluoropyridine-3-carboxaldehyde, pubchem5150, zlchem 1162, acmc-1b2mv
C1=CC(=NC=C1C=O)F
125.1
125.1
Glass bottle
677728-92-6
C6H4FNO
PZPNGWWKCSJKOS-UHFFFAOYSA-N
6-fluoropyridine-3-carbaldehyde
16414246
95%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 6-Fluoropyridine-3-carboxaldehyde

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documents