Learn More
6-Chloro-3-pyridinol, 97%, Thermo Scientific™
Catalog Number p-7077254
Click to view available options
Quantity:
1 g
5 g
41288-96-4
C5H4ClNO
129.54
MFCD00234050
KVCOOWROABTXDJ-UHFFFAOYSA-N
2-chloro-5-hydroxypyridine, 6-chloro-3-pyridinol, 3-pyridinol, 6-chloro, 2-chloro-5-hydroxy pyridine, 2-chloro-5-pyridinol, 2-chloropyridin-5-ol, pubchem2410, zlchem 1336, 6-chloropyridine-3-ol, 6-chloro-pyridin-3-ol
819821
6-chloropyridin-3-ol
OC1=CC=C(Cl)N=C1
41288-96-4
C5H4ClNO
129.54
MFCD00234050
KVCOOWROABTXDJ-UHFFFAOYSA-N
2-chloro-5-hydroxypyridine, 6-chloro-3-pyridinol, 3-pyridinol, 6-chloro, 2-chloro-5-hydroxy pyridine, 2-chloro-5-pyridinol, 2-chloropyridin-5-ol, pubchem2410, zlchem 1336, 6-chloropyridine-3-ol, 6-chloro-pyridin-3-ol
819821
6-chloropyridin-3-ol
OC1=CC=C(Cl)N=C1
Melting Point 159.0°C to 161.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 129.54
Percent Purity 97%
Chemical Name or Material 6-Chloro-3-pyridinol
compliance-icons
Product Identifier
  • 6-Chloro-3-pyridinol, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.