missing translation for 'onlineSavingsMsg'
Learn More

6-Aza-2-thiothymine, 98%, Thermo Scientific Chemicals

Catalog Number p-7021714
Click to view available options
Quantity:
5 g
25 g
100 g
615-76-9
C4H5N3OS
143.164
MFCD00006458
NKOPQOSBROLOFP-UHFFFAOYSA-N
6-aza-2-thiothymine, 2-thio-6-azathymine, 6-azathiothymine, 6-methyl-3-thioxo-3,4-dihydro-1,2,4-triazin-5 2h-one, 3-mercapto-6-methyl-1,2,4-triazin-5 2h-one, 1,2,4-triazin-5 2h-one, 3,4-dihydro-6-methyl-3-thioxo, 3-mercapto-6-methyl-as-triazin-5-ol, as-triazin-5-ol, 3-mercapto-6-methyl, 6-methyl-3-sulfanyl-1,2,4-triazin-5-ol, 3,4-dihydro-6-methyl-3-thioxo-1,2,4-triazin-5 2h-one
1263666
6-methyl-3-sulfanylidene-2H-1,2,4-triazin-5-one
CC1=NNC(=S)NC1=O
615-76-9
C4H5N3OS
143.164
MFCD00006458
NKOPQOSBROLOFP-UHFFFAOYSA-N
6-aza-2-thiothymine, 2-thio-6-azathymine, 6-azathiothymine, 6-methyl-3-thioxo-3,4-dihydro-1,2,4-triazin-5 2h-one, 3-mercapto-6-methyl-1,2,4-triazin-5 2h-one, 1,2,4-triazin-5 2h-one, 3,4-dihydro-6-methyl-3-thioxo, 3-mercapto-6-methyl-as-triazin-5-ol, as-triazin-5-ol, 3-mercapto-6-methyl, 6-methyl-3-sulfanyl-1,2,4-triazin-5-ol, 3,4-dihydro-6-methyl-3-thioxo-1,2,4-triazin-5 2h-one
1263666
6-methyl-3-sulfanylidene-2H-1,2,4-triazin-5-one
CC1=NNC(=S)NC1=O

6-Aza-2-thiothymine is used in MALDI analysis of acidic glycans in negative ion mode. It is also called as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Aza-2-thiothymine is used in MALDI analysis of acidic glycans in negative ion mode. It is also called as pharmaceutical intermediates.

Solubility
Soluble in water (partly), and formic acid (5 %).

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY
Melting Point 218°C to 221°C
Quantity 5 g
Beilstein 4861
Solubility Information Soluble in water (partly),and formic acid (5 %).
Formula Weight 143.17
Percent Purity 98%
Chemical Name or Material 6-Aza-2-thiothymine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'