missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Aminocoumarin hydrochloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
6-Aminocoumarin hydrochloride is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Aminocoumarin hydrochloride is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
6-Aminocoumarin hydrochloride is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Specifications
Melting Point | >260°C (decomposition) |
Quantity | 1 g |
Beilstein | 3698955 |
Solubility Information | Insoluble in water. |
Formula Weight | 197.62 |
Percent Purity | 97% |
Chemical Name or Material | 6-Aminocoumarin hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'