missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroquinazolin-4-ol, 97%, Thermo Scientific™

Catalog Number p-2702834
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
436-72-6
C8H5FN2O
164.14
MFCD00173674
UXEZULVIMJVIFB-UHFFFAOYSA-N
5-fluoro-4-hydroxyquinazoline, 5-fluoroquinazolin-4-ol, 5-fluoroquinazolin-4 1h-one, 5-fluoroquinazolin-4 3h-one, 5-fluoro-4-quinazolone, 5-fluoro-3h-quinazolin-4-one, 4 1h-quinazolinone, 5-fluoro, pubchem6307, 5-fluoroquinazolin-4-one, acmc-209jv3
2737331
FC1=C2C(NC=NC2=O)=CC=C1
This item is not returnable. View return policy
436-72-6
C8H5FN2O
164.14
MFCD00173674
UXEZULVIMJVIFB-UHFFFAOYSA-N
5-fluoro-4-hydroxyquinazoline, 5-fluoroquinazolin-4-ol, 5-fluoroquinazolin-4 1h-one, 5-fluoroquinazolin-4 3h-one, 5-fluoro-4-quinazolone, 5-fluoro-3h-quinazolin-4-one, 4 1h-quinazolinone, 5-fluoro, pubchem6307, 5-fluoroquinazolin-4-one, acmc-209jv3
2737331
FC1=C2C(NC=NC2=O)=CC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 5-fluoro-1,4-dihydroquinazolin-4-one
Formula Weight 164.14
Percent Purity 97%
Chemical Name or Material 5-Fluoroquinazolin-4-ol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'