missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-iodobenzoic acid, 97%, Thermo Scientific™

Catalog Number 15457438
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 52548-63-7
Melting Point 145°C to 149°C
Molecular Formula C7H4FIO2
MDL Number MFCD00837316
Quantity 1 g
UN Number UN2811
Sensitivity Light sensitive
Synonym 2-iodo-5-fluorobenzoic acid, benzoic acid, 5-fluoro-2-iodo, 5-fluoro-2-iodo-benzoic acid, pubchem1391, acmc-209zxz, akos akm00947, 3-fluoro-6-iodo benzoic acid, benzoic acid,5-fluoro-2-iodo, 5-fluoro-2-iodobenzoic acid
InChI Key XPFMQYOPTHMSJJ-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1F)C(=O)O)I
IUPAC Name 5-fluoro-2-iodobenzoic acid
Molecular Weight (g/mol) 266.01
PubChem CID 13158583
Formula Weight 266.01
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-iodobenzoic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'