missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-3-methyl-1-benzothiophene-2-carboxylic acid, Technical Grade, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 226.68 |
Percent Purity | 95% |
Chemical Name or Material | 5-Chloro-3-methyl-1-benzothiophene-2-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'