missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-fluorobenzylamine, 97%, Thermo Scientific™

Catalog Number p-7047410
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
261723-26-6
C7H7ClFN
159.59
MFCD01631440
BFLBLJKULFDOTN-UHFFFAOYSA-N
5-chloro-2-fluorobenzylamine, 5-chloro-2-fluorophenyl methanamine, 2-fluoro-5-chlorobenzylamine, 2-fluoro-5-chlorobenzyl amine, 1-5-chloro-2-fluorophenyl methanamine, 5-chloro-2-fluoro-benzylamine, 5-chloro-2-fluorophenyl methylamine, 5-chloro-2-fluorobenzyl amine, pubchem4378
2773616
NCC1=CC(Cl)=CC=C1F
This item is not returnable. View return policy
261723-26-6
C7H7ClFN
159.59
MFCD01631440
BFLBLJKULFDOTN-UHFFFAOYSA-N
5-chloro-2-fluorobenzylamine, 5-chloro-2-fluorophenyl methanamine, 2-fluoro-5-chlorobenzylamine, 2-fluoro-5-chlorobenzyl amine, 1-5-chloro-2-fluorophenyl methanamine, 5-chloro-2-fluoro-benzylamine, 5-chloro-2-fluorophenyl methylamine, 5-chloro-2-fluorobenzyl amine, pubchem4378
2773616
NCC1=CC(Cl)=CC=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.536
Quantity 250 mg
UN Number UN2735
Sensitivity Air Sensitive
IUPAC Name (5-chloro-2-fluorophenyl)methanamine
Formula Weight 159.59
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-fluorobenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'