missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-Chloro-2-fluorobenzoic acid, 97%
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Melting Point | 148°C to 150°C |
Quantity | 1 g |
Beilstein | 2614286 |
Formula Weight | 174.56 |
Percent Purity | 97% |
Chemical Name or Material | 5-Chloro-2-fluorobenzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'