missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-cyanobenzeneboronic acid, 98%, Thermo Scientific™

Catalog Number p-7054004
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1072946-52-1
C7H5BClNO2
181.382
JXIINQQPEHZQAY-UHFFFAOYSA-N
5-chloro-2-cyanophenyl boronic acid, 5-chloro-2-cyanophenyl boronicacid, boronicacid,b-5-chloro-2-cyanophenyl
46738934
(5-chloro-2-cyanophenyl)boronic acid
B(C1=C(C=CC(=C1)Cl)C#N)(O)O
This item is not returnable. View return policy
1072946-52-1
C7H5BClNO2
181.382
JXIINQQPEHZQAY-UHFFFAOYSA-N
5-chloro-2-cyanophenyl boronic acid, 5-chloro-2-cyanophenyl boronicacid, boronicacid,b-5-chloro-2-cyanophenyl
46738934
(5-chloro-2-cyanophenyl)boronic acid
B(C1=C(C=CC(=C1)Cl)C#N)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 225.84
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Chloro-2-cyanobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'