missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-2-fluorobenzamide, 96%

Brand:  Alfa Aesar™ H62457.03

Additional Details : CAS Number : 214210-17-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458968

  • 376.00 EUR / 1g


Estimated Shipment date
from Supplier 21-02-2019
Add to basket

Specifications

Chemical Identifiers

214210-17-0
218.025
MAGQVUXQRXLNGB-UHFFFAOYSA-N
2778901
C1=CC(=C(C=C1Br)C(=O)N)F
C7H5BrFNO
MFCD03094430
5-bromo-2-fluoro-benzamide, 2-fluoro-5-bromobenzamide
5-bromo-2-fluorobenzamide

Specifications

5-Bromo-2-fluorobenzamide
214210-17-0
MFCD03094430
MAGQVUXQRXLNGB-UHFFFAOYSA-N
5-bromo-2-fluorobenzamide
2778901
Solid
1g
C7H5BrFNO
5-bromo-2-fluoro-benzamide, 2-fluoro-5-bromobenzamide
C1=CC(=C(C=C1Br)C(=O)N)F
218.025
218.02
96%
Videos
SDS