missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2'-Deoxyuridine, MP Biomedicals™

Catalog Number p-7083098
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy
59-14-3
C9H11BrN2O5
307.10
MFCD00006529
WOVKYSAHUYNSMH-RRKCRQDMSA-N
5-bromo-2'-deoxyuridine, broxuridine, bromodeoxyuridine, brdu, 5-bromodeoxyuridine, 5-brdu, budr, 5-bromouracil deoxyriboside, bromouracil deoxyriboside, 5-bromodesoxyuridine
6035
CHEBI:472552
5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O
This item is not returnable. View return policy
59-14-3
C9H11BrN2O5
307.10
MFCD00006529
WOVKYSAHUYNSMH-RRKCRQDMSA-N
5-bromo-2'-deoxyuridine, broxuridine, bromodeoxyuridine, brdu, 5-bromodeoxyuridine, 5-brdu, budr, 5-bromouracil deoxyriboside, bromouracil deoxyriboside, 5-bromodesoxyuridine
6035
CHEBI:472552
5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O

Useful for the study of DNA synthesis. 5-Bromo-2'-Deoxyuridine, MP Biomedicals (5-BrdU) is a thymidine analogue that is incorporated into DNA. It is routinely and extensively used to measure DNA synthesis and to label dividing cells. Consequently it is us

5-BrdU can be used to label DNA, which can be used in conjunction with BrdU Monoclonal Antibody for the immunohistochemical analysis of cell proliferation. Thymidine analog is used as a mutagen in genetic research. It is selectively incorporated into cellular DNA in place of thymidine during S-phase. It is also used to study cell signaling and other processes that induce cell proliferation.

Melting Point 183°C to 193°C
Quantity 250 mg
Solubility Information Soluble in water (10mg/mL or 50–100mg/mL with heat) or sodium hydroxide (50mg/mL).
Formula Weight 307.1
Percent Purity ≥99%
Physical Form Powder
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'