missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Vinylphenol, min 10% soln. in propylene glycol, Thermo Scientific Chemicals
32.26 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
4-Vinylphenol acts as a monomer and used in the preparation of poly-4-vinylphenol polymer, which is used in electronics as a dielectric layer in organic transistors in organic thin-film-transistor liquid-crystal displays (TFT LCD). It is a phenolic compound and presents in wine and beer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Vinylphenol acts as a monomer and used in the preparation of poly-4-vinylphenol polymer, which is used in electronics as a dielectric layer in organic transistors in organic thin-film-transistor liquid-crystal displays (TFT LCD). It is a phenolic compound and presents in wine and beer.
Solubility
Slightly miscible with water.
Notes
Hygroscopic. Moisture and heat sensitive. Incompatible with oxidizing agents.
4-Vinylphenol acts as a monomer and used in the preparation of poly-4-vinylphenol polymer, which is used in electronics as a dielectric layer in organic transistors in organic thin-film-transistor liquid-crystal displays (TFT LCD). It is a phenolic compound and presents in wine and beer.
Solubility
Slightly miscible with water.
Notes
Hygroscopic. Moisture and heat sensitive. Incompatible with oxidizing agents.
Specifications
Density | 1.04 |
Flash Point | >100°C (212°F) |
Refractive Index | 1.447 |
Quantity | 1 g |
Beilstein | 506844 |
Sensitivity | Hygroscopic |
Solubility Information | Slightly miscible with water. |
Formula Weight | 120.15 |
Concentration or Composition (by Analyte or Components) | ≥10% soln. in propylene glycol |
Chemical Name or Material | 4-Vinylphenol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'