Learn More
4-(Trifluoromethyl)styrene, 98%, stab., Thermo Scientific Chemicals
Catalog Number p-7035786
Click to view available options
Quantity:
1 g
5 g
402-50-6
C9H7F3
172.15
MFCD00075539
CEWDRCQPGANDRS-UHFFFAOYSA-N
1-trifluoromethyl-4-vinylbenzene, 4-trifluoromethyl styrene, 4-trifluoromethylstyrene, 1-ethenyl-4-trifluoromethyl benzene, 4-vinylbenzotrifluoride, 1-trifluoromethyl-4-vinyl-benzene, 4-trifluoromethystyrene, p-trifluoromethylstyrene, pubchem15485, acmc-1aivr
2782798
1-ethenyl-4-(trifluoromethyl)benzene
C=CC1=CC=C(C=C1)C(F)(F)F
402-50-6
C9H7F3
172.15
MFCD00075539
CEWDRCQPGANDRS-UHFFFAOYSA-N
1-trifluoromethyl-4-vinylbenzene, 4-trifluoromethyl styrene, 4-trifluoromethylstyrene, 1-ethenyl-4-trifluoromethyl benzene, 4-vinylbenzotrifluoride, 1-trifluoromethyl-4-vinyl-benzene, 4-trifluoromethystyrene, p-trifluoromethylstyrene, pubchem15485, acmc-1aivr
2782798
1-ethenyl-4-(trifluoromethyl)benzene
C=CC1=CC=C(C=C1)C(F)(F)F
4-(Trifluoromethyl)styrene is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Trifluoromethyl)styrene is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Moisture & light sensitive. Incompatible with oxidizing agents, acids, bases, heat, ultravioloet radiation and free radical initiators.
TRUSTED_SUSTAINABILITY
Density 1.165
Boiling Point 65°C to 66°C (40 mmHg)
Flash Point 42°C (108°F)
Refractive Index 1.466
Quantity 1 g
UN Number UN1993
Beilstein 1863548
Sensitivity Light and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 172.15
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethyl)styrene, stabilized