missing translation for 'onlineSavingsMsg'
Learn More

(4-Pyrid-2-ylphenyl)methanol, ≥97%, Maybridge™

Brand:  Maybridge CC41509DA

Additional Details : CAS Number : 98061-39-3 Weight : 0.05100kg

Quantity 1g

 View more versions of this product

Product Code. 10022364

  • 145.60 EUR / 1g

Estimated Shipment: 22-09-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

98061-39-3
185.226
BESAKUXOHCFPAA-UHFFFAOYSA-N
1515239
C1=CC=NC(=C1)C2=CC=C(C=C2)CO
C12H11NO
MFCD04116292
2-4-hydroxymethylphenyl pyridine, 4-pyridin-2-yl phenyl methanol, 4-pyrid-2-ylphenyl methanol, 4-pyridin-2-yl benzyl alcohol, 4-2-pyridyl benzyl alcohol, 4-2-pyridinyl benzylalcohol, 4-2-pyridinyl benzyl alcohol, 4-pyrid-2-yl benzyl alcohol, 4-2-pyridyl phenyl methanol
(4-pyridin-2-ylphenyl)methanol

Specifications

(4-Pyrid-2-ylphenyl)methanol
1g
96.5
97%
MFCD04116292
BESAKUXOHCFPAA-UHFFFAOYSA-N
(4-pyridin-2-ylphenyl)methanol
1515239
97%
Amber Glass Bottle
98061-39-3
100.0
C12H11NO
2-4-hydroxymethylphenyl pyridine, 4-pyridin-2-yl phenyl methanol, 4-pyrid-2-ylphenyl methanol, 4-pyridin-2-yl benzyl alcohol, 4-2-pyridyl benzyl alcohol, 4-2-pyridinyl benzylalcohol, 4-2-pyridinyl benzyl alcohol, 4-pyrid-2-yl benzyl alcohol, 4-2-pyridyl phenyl methanol
C1=CC=NC(=C1)C2=CC=C(C=C2)CO
185.226
185.22
Videos
SDS Documents
Certificates