missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrostyrene, 98%, Thermo Scientific™

Catalog Number p-7030533
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
100-13-0
C8H7NO2
149.15
MFCD00041254
YFZHODLXYNDBSM-UHFFFAOYSA-N
4-nitrostyrene, 1-nitro-4-vinylbenzene, p-nitrostyrene, benzene, 1-ethenyl-4-nitro, ccris 1674, styrene, p-nitro, benzene, 1-ethenyl-4-nitro-9ci, nitrovinylbenzene, 4-nitro-styrene, p-nitrophenylethylene
7481
1-ethenyl-4-nitrobenzene
[O-][N+](=O)C1=CC=C(C=C)C=C1
This item is not returnable. View return policy
100-13-0
C8H7NO2
149.15
MFCD00041254
YFZHODLXYNDBSM-UHFFFAOYSA-N
4-nitrostyrene, 1-nitro-4-vinylbenzene, p-nitrostyrene, benzene, 1-ethenyl-4-nitro, ccris 1674, styrene, p-nitro, benzene, 1-ethenyl-4-nitro-9ci, nitrovinylbenzene, 4-nitro-styrene, p-nitrophenylethylene
7481
1-ethenyl-4-nitrobenzene
[O-][N+](=O)C1=CC=C(C=C)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Can be used to introduce amino groups into polymers. The thermal degradation of styrene-4-nitrostyrene copolymers (SNS) has been studied using differential thermal analysis (DTA) and thermogravimetry (TGA) under isothermal and dynamic conditions in dynamic nitrogen. 4—Nitrostyrene gives isomeric (4-nitropheny1)-propiona1dehydes in a 96% yield.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, air, heat. Store at 4 deg C.
TRUSTED_SUSTAINABILITY
Melting Point 25°C to 28°C
Density 1.163
Boiling Point 93°C to 96°C (3 mmHg)
Flash Point 109°C (228°F)
Refractive Index 1.6045
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 149.15
Percent Purity 98%
Chemical Name or Material 4-Nitrostyrene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'