missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrobenzo-15-crown-5, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-Nitrobenzo-15-crown-5 is a raw material used in synthesis as well as an intemediate in chemical syntheis and pharmaceutica intemediate. It is also used to separate cations, dissolve inorganic salts into organic solvents to perform naked anion chemistry, serve as phase transfer catalysts, and chiral crown ethers have been used to resolve racemic mixtures or separate isomers by chiral chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrobenzo-15-crown-5 is a raw material used in synthesis as well as an intemediate in chemical syntheis and pharmaceutica intemediate. It is also used to separate cations, dissolve inorganic salts into organic solvents to perform naked anion chemistry, serve as phase transfer catalysts, and chiral crown ethers have been used to resolve racemic mixtures or separate isomers by chiral chromatography.
Solubility
It is soluble in non-polar solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases
4-Nitrobenzo-15-crown-5 is a raw material used in synthesis as well as an intemediate in chemical syntheis and pharmaceutica intemediate. It is also used to separate cations, dissolve inorganic salts into organic solvents to perform naked anion chemistry, serve as phase transfer catalysts, and chiral crown ethers have been used to resolve racemic mixtures or separate isomers by chiral chromatography.
Solubility
It is soluble in non-polar solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases
Specifications
Melting Point | 93°C to 95°C |
Quantity | 1 g |
Solubility Information | It is soluble in non-polar solvents. |
Formula Weight | 313.3 |
Percent Purity | 99% |
Chemical Name or Material | 4-Nitrobenzo-15-crown-5 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'