missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylbenzyl alcohol, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
4-Methylbenzyl alcohol is used in fragrances, flavoring and as a laboratory reagent. It is also used as a raw material for the preparation of polycarbonates and 4-methyl-benzaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methylbenzyl alcohol is used in fragrances, flavoring and as a laboratory reagent. It is also used as a raw material for the preparation of polycarbonates and 4-methyl-benzaldehyde.
Solubility
Soluble in methanol, ether and ethanol. Slightly soluble in water.
Notes
Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
4-Methylbenzyl alcohol is used in fragrances, flavoring and as a laboratory reagent. It is also used as a raw material for the preparation of polycarbonates and 4-methyl-benzaldehyde.
Solubility
Soluble in methanol, ether and ethanol. Slightly soluble in water.
Notes
Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
Specifications
Melting Point | 57°C to 61°C |
Density | 0.978 |
Boiling Point | 217°C |
Quantity | 25 g |
Beilstein | 1856550 |
Solubility Information | Soluble in methanol,ether and ethanol. Slightly soluble in water. |
Formula Weight | 122.17 |
Percent Purity | 99% |
Chemical Name or Material | 4-Methylbenzyl alcohol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'