missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzylamine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7022292
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
2393-23-9
C8H11NO
137.182
MFCD00008122
IDPURXSQCKYKIJ-UHFFFAOYSA-N
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
75452
CHEBI:49837
(4-methoxyphenyl)methanamine
COC1=CC=C(C=C1)CN
This item is not returnable. View return policy
2393-23-9
C8H11NO
137.182
MFCD00008122
IDPURXSQCKYKIJ-UHFFFAOYSA-N
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
75452
CHEBI:49837
(4-methoxyphenyl)methanamine
COC1=CC=C(C=C1)CN

4-Methoxybenzylamine is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used fine chemical intemediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzylamine is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used fine chemical intemediate.

Solubility
It is highly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Store under inert gas. Keep away from Strong oxidizing agentsacids, Acid chlorides, Acid anhydrides, Oxidizing agents, Chloroformates.
TRUSTED_SUSTAINABILITY
Melting Point -10°C
Density 1.053
Boiling Point 236°C to 237°C
Flash Point 114°C (237°F)
Odor Amine-like
Refractive Index 1.546
Quantity 25 g
UN Number UN2735
Beilstein 508206
Sensitivity Air sensitive; May form a precipitate
Solubility Information It is highly soluble in water.
Formula Weight 137.18
Percent Purity ≥98%
Chemical Name or Material 4-Methoxybenzylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'