missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methoxy-N-(2,4-dimethylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59310.03

Additional Details : CAS Number : 127291-95-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15485578

  • 122.60 EUR / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

4-Methoxy-N-(2,4-dimethylphenyl)benzamide
255.32
3551313
127291-95-6
MFCD00425357

Specifications

4-Methoxy-N-(2,4-dimethylphenyl)benzamide
127291-95-6
97%
MFCD00425357
1g
Solid
255.32
3551313
Videos
SDS