missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4813130
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1798-06-7
C8H7IO2
262.05
FJSHTWVDFAUNCO-UHFFFAOYSA-N
4-iodophenylacetic acid, 2-4-iodophenyl acetic acid, 4-iodophenyl acetic acid, 4-iodophenylaceticacid, p-iodophenylacetic acid, benzeneacetic acid, 4-iodo, 4-iodo-phenyl-acetic acid, pubchem18835, 4-iodoohenylacetic acid, 4-iodobenzeneacetic acid
137214
2-(4-iodophenyl)acetic acid
C1=CC(=CC=C1CC(=O)O)I
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1798-06-7
C8H7IO2
262.05
FJSHTWVDFAUNCO-UHFFFAOYSA-N
4-iodophenylacetic acid, 2-4-iodophenyl acetic acid, 4-iodophenyl acetic acid, 4-iodophenylaceticacid, p-iodophenylacetic acid, benzeneacetic acid, 4-iodo, 4-iodo-phenyl-acetic acid, pubchem18835, 4-iodoohenylacetic acid, 4-iodobenzeneacetic acid
137214
2-(4-iodophenyl)acetic acid
C1=CC(=CC=C1CC(=O)O)I

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 140°C to 141.5°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 262.05
Percent Purity 97%
Chemical Name or Material 4-Iodophenylacetic acid
compliance-icons
Product Identifier
  • 4-Iodophenylacetic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'