missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4'-Hydroxybiphenyl-4-carboxamidoxime, 97%

Brand:  Alfa Aesar™ H58802.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15445008

  • 414.00 EUR / 1g


Estimated Shipment date
from Supplier 19-06-2019
Add to basket

Specifications

Chemical Identifiers

885952-07-8
228.25
HQIIILHMSQHPJV-UHFFFAOYSA-N
6002849
C13H12N2O2
MFCD22581245
4'-hydroxybiphenyl-4-carboxamidoxime, n,4'-dihydroxy-biphenyl-4-carboximidamide, n,4'-dihydroxy-1,1'-biphenyl-4-carboximidamide, 4-4-amino-hydroxyamino methylidene cyclohexa-2,5-dien-1-ylidene cyclohexa-2,5-dien-1-one

Specifications

4'-Hydroxybiphenyl-4-carboxamidoxime
1g
C13H12N2O2
4'-hydroxybiphenyl-4-carboxamidoxime, n,4'-dihydroxy-biphenyl-4-carboximidamide, n,4'-dihydroxy-1,1'-biphenyl-4-carboximidamide, 4-4-amino-hydroxyamino methylidene cyclohexa-2,5-dien-1-ylidene cyclohexa-2,5-dien-1-one
228.25
228.25
97%
Brown
885952-07-8
MFCD22581245
HQIIILHMSQHPJV-UHFFFAOYSA-N
6002849
Solid
Videos
SDS