missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxybenzyl alcohol, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
100 g
250 g
Description
4-Hydroxybenzyl alcohol is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxybenzyl alcohol is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water (6.7 mg/ml at 20°C), dioxane (100 mg/ml), 1N NaOH (50 mg/ml), DMSO, and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acid chlorides, acid anhydrides, bases, oxidizing agents.
4-Hydroxybenzyl alcohol is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water (6.7 mg/ml at 20°C), dioxane (100 mg/ml), 1N NaOH (50 mg/ml), DMSO, and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acid chlorides, acid anhydrides, bases, oxidizing agents.
Specifications
Melting Point | 114°C to 122°C |
Boiling Point | 251°C to 253°C |
Quantity | 50 g |
Beilstein | 1858967 |
Sensitivity | Air and light sensitive |
Solubility Information | Soluble in water (6.7mg/ml at 20°C),dioxane (100mg/ml),1N NaOH (50mg/ml),DMSO,and methanol. |
Formula Weight | 124.14 |
Percent Purity | 99% |
Chemical Name or Material | 4-Hydroxybenzyl alcohol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'