missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Hydroxy-2-methyl-6-(trifluoromethyl)pyrimidine, 95%

Brand:  Alfa Aesar™ H64364.MD

Additional Details : Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15458876

  • 65.00 EUR / 250mg


Estimated Shipment date
from Supplier 18-06-2019
Add to basket

Specifications

Chemical Identifiers

2836-44-4
178.114
DBDRDWKBAGNHHS-UHFFFAOYSA-N
233901
CC1=NC(=O)C=C(N1)C(F)(F)F
C6H5F3N2O
MFCD03265321
2-methyl-6-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-methyl-6-trifluoromethylpyrimidine, 4-hydroxy-2-methyl-6-trifluoromethyl pyrimidine, 4-pyrimidinol, 2-methyl-6-trifluoromethyl, 2-methyl-6-trifluoromethylpyrimidin-4-ol, 2-methyl-6-trifluoromethyl-4-pyrimidinol, 2-methyl-4-hydroxy-6-trifluoromethylpyrimidine, 2-methyl-6-trifluoromethyl-1h-pyrimidin-4-one, 2-methyl-6-trifluoromethyl pyrimidin-4 3h-one, 4 1h-pyrimidinone, 2-methyl-6-trifluoromethyl
2-methyl-6-(trifluoromethyl)-1H-pyrimidin-4-one

Specifications

4-Hydroxy-2-methyl-6-(trifluoromethyl)pyrimidine
2836-44-4
MFCD03265321
DBDRDWKBAGNHHS-UHFFFAOYSA-N
2-methyl-6-(trifluoromethyl)-1H-pyrimidin-4-one
233901
Solid
250mg
C6H5F3N2O
2-methyl-6-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-methyl-6-trifluoromethylpyrimidine, 4-hydroxy-2-methyl-6-trifluoromethyl pyrimidine, 4-pyrimidinol, 2-methyl-6-trifluoromethyl, 2-methyl-6-trifluoromethylpyrimidin-4-ol, 2-methyl-6-trifluoromethyl-4-pyrimidinol, 2-methyl-4-hydroxy-6-trifluoromethylpyrimidine, 2-methyl-6-trifluoromethyl-1h-pyrimidin-4-one, 2-methyl-6-trifluoromethyl pyrimidin-4 3h-one, 4 1h-pyrimidinone, 2-methyl-6-trifluoromethyl
CC1=NC(=O)C=C(N1)C(F)(F)F
178.114
178.11
95%
Videos
SDS