missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorostyrene, 97+%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals
25.13 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Description
4-Fluorostyrene, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Fluorostyrene, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from acids, bases, oxidizing agents, halogens.
4-Fluorostyrene, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from acids, bases, oxidizing agents, halogens.
Specifications
Melting Point | -36°C |
Density | 1.024 |
Boiling Point | 145°C to 148°C |
Flash Point | 26°C (78°F) |
Odor | Recognizable |
Refractive Index | 1.515 |
Quantity | 1 g |
UN Number | UN1993 |
Beilstein | 1634203 |
Solubility Information | Insoluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'