missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Fluoro-3-(methoxycarbamoyl)benzeneboronic acid, 98%

Catalog Number p-7035161
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
913835-47-9
C8H9BFNO4
212.971
MFCD08235105
OJNRHYRWHLOKBF-UHFFFAOYSA-N
n-methoxy 5-borono-2-fluorobenzamide, 4-fluoro-3-methoxycarbamoyl phenyl boronic acid, 4-fluoro-3-methoxyamino carbonyl benzeneboronic acid, 4-fluoro-3-methoxycarbamoyl phenylboronic acid, acmc-209r9d, 4-fluoro-3-methoxycarbamoyl phenyl boronicacid, 4-fluoranyl-3-methoxycarbamoyl phenyl boronic acid, 4-fluoro-3-methoxyamino-oxomethyl phenyl boronic acid
44717602
[4-fluoro-3-(methoxycarbamoyl)phenyl]boronic acid
B(C1=CC(=C(C=C1)F)C(=O)NOC)(O)O
This item is not returnable. View return policy
913835-47-9
C8H9BFNO4
212.971
MFCD08235105
OJNRHYRWHLOKBF-UHFFFAOYSA-N
n-methoxy 5-borono-2-fluorobenzamide, 4-fluoro-3-methoxycarbamoyl phenyl boronic acid, 4-fluoro-3-methoxyamino carbonyl benzeneboronic acid, 4-fluoro-3-methoxycarbamoyl phenylboronic acid, acmc-209r9d, 4-fluoro-3-methoxycarbamoyl phenyl boronicacid, 4-fluoranyl-3-methoxycarbamoyl phenyl boronic acid, 4-fluoro-3-methoxyamino-oxomethyl phenyl boronic acid
44717602
[4-fluoro-3-(methoxycarbamoyl)phenyl]boronic acid
B(C1=CC(=C(C=C1)F)C(=O)NOC)(O)O
Melting Point 213°C to 215°C
Quantity 250 mg
Formula Weight 212.97
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-(methoxycarbamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'