missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-2,6-difluorobenzamide, 97%, Thermo Scientific™

Catalog Number 15434837
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 916483-57-3
Melting Point 187°C to 191°C
Molecular Formula C9H9F2NO2
MDL Number MFCD09832395
Quantity 250 mg
Synonym benzamide, 4-ethoxy-2,6-difluoro, 4-ethoxy-2,6-difluorobenzamide, jrd
InChI Key YLNGDTJVZFEUJH-UHFFFAOYSA-N
SMILES CCOC1=CC(F)=C(C(N)=O)C(F)=C1
IUPAC Name 4-ethoxy-2,6-difluorobenzamide
Molecular Weight (g/mol) 201.17
PubChem CID 46737698
Formula Weight 201.17
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-2,6-difluorobenzamide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'