missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorobenzyl)triphenylphosphonium chloride, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081691
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
1530-39-8
C25H21Cl2P
423.32
MFCD00041533
RAHOAHBOOHXRDY-UHFFFAOYSA-M
4-chlorobenzyl triphenylphosphonium chloride, 4-chlorobenzyl triphenylphosphoniumchloride, 4-chlorophenyl methyl triphenylphosphanium chloride, acmc-1bpq8, 4-chlorobenzyltriphenylphosphonium chloride, 4-chlorobenzyltriphenyl-phosphonium chloride, 4-chlorophenyl methyl-triphenylphosphanium chloride, 4-chlorophenyl methyl-triphenylphosphonium chloride, 4-chlorophenyl methyl triphenylphosphine, chloride, 4-chlorobenzyl triphenyl phosphonium chloride
2733546
[Cl-].ClC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
This item is not returnable. View return policy
1530-39-8
C25H21Cl2P
423.32
MFCD00041533
RAHOAHBOOHXRDY-UHFFFAOYSA-M
4-chlorobenzyl triphenylphosphonium chloride, 4-chlorobenzyl triphenylphosphoniumchloride, 4-chlorophenyl methyl triphenylphosphanium chloride, acmc-1bpq8, 4-chlorobenzyltriphenylphosphonium chloride, 4-chlorobenzyltriphenyl-phosphonium chloride, 4-chlorophenyl methyl-triphenylphosphanium chloride, 4-chlorophenyl methyl-triphenylphosphonium chloride, 4-chlorophenyl methyl triphenylphosphine, chloride, 4-chlorobenzyl triphenyl phosphonium chloride
2733546
[Cl-].ClC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 286°C to 291°C
Quantity 25 g
Beilstein 4173400
Sensitivity Hygroscopic
IUPAC Name (4-chlorophenyl)methyl-triphenylphosphanium;chloride
Formula Weight 423.32
Percent Purity ≥98%
Chemical Name or Material (4-Chlorobenzyl)triphenylphosphonium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'