missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorobenzylamine, 97%, Thermo Scientific™

Catalog Number p-7029330
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
72235-58-6
C7H7ClFN
159.588
MFCD04116008
HSNPBYKCCNMQNA-UHFFFAOYSA-N
4-chloro-3-fluorophenyl methanamine, 4-chloro-3-fluorobenzylamine, 4-chloro-3-fluorobenzyl amine, benzenemethanamine,4-chloro-3-fluoro, benzenemethanamine, 4-chloro-3-fluoro, 4-chloro-3-fluorophenyl methylamine, 1-4-chloro-3-fluorophenyl methanamine, 3-fluoro-4-chlorobenzyl amine, rarechem al bw 1597, 4-chloranyl-3-fluoranyl-phenyl methanamine
2757541
(4-chloro-3-fluorophenyl)methanamine
C1=CC(=C(C=C1CN)F)Cl
This item is not returnable. View return policy
72235-58-6
C7H7ClFN
159.588
MFCD04116008
HSNPBYKCCNMQNA-UHFFFAOYSA-N
4-chloro-3-fluorophenyl methanamine, 4-chloro-3-fluorobenzylamine, 4-chloro-3-fluorobenzyl amine, benzenemethanamine,4-chloro-3-fluoro, benzenemethanamine, 4-chloro-3-fluoro, 4-chloro-3-fluorophenyl methylamine, 1-4-chloro-3-fluorophenyl methanamine, 3-fluoro-4-chlorobenzyl amine, rarechem al bw 1597, 4-chloranyl-3-fluoranyl-phenyl methanamine
2757541
(4-chloro-3-fluorophenyl)methanamine
C1=CC(=C(C=C1CN)F)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 159.59
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-fluorobenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'