missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-1-methyl-1H-pyrazole-3-carbaldehyde, 95+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
IUPAC Name | 4-chloro-1-methyl-1H-pyrazole-3-carbaldehyde |
Formula Weight | 144.56 |
Percent Purity | 95% |
Chemical Name or Material | 4-Chloro-1-methyl-1H-pyrazole-3-carbaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'